Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MSK1 (Ser376) Recombinant Rabbit Monoclonal Antibody (2F3)
GREENER_CHOICE

Catalog No. FEMA549768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549768 Supplier Invitrogen™ Supplier No. MA549768
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-MSK1 (Ser376) Recombinant Monoclonal Antibody for IHC (P), ELISA

RPS6KA5 (MSK1) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes. RPS6KA5 is required for the mitogen or stress-induced phosphorylation of the transcription factors CREB1 and ATF1 and for the regulation of the transcription factors RELA, STAT3 and ETV1/ER81, and that contributes to gene activation by histone phosphorylation and functions in the regulation of inflammatory genes. Phosphorylates CREB1 and ATF1 in response to mitogenic or stress stimuli such as UV-C irradiation, epidermal growth factor (EGF) and anisomycin. Plays an essential role in the control of RELA transcriptional activity in response to TNF and upon glucocorticoid, associates in the cytoplasm with the glucocorticoid receptor NR3C1 and contributes to RELA inhibition and repression of inflammatory gene expression. In skeletal myoblasts is required for phosphorylation of RELA at 'Ser-276' during oxidative stress. In erythropoietin-stimulated cells, is necessary for the 'Ser-727' phosphorylation of STAT3 and regulation of its transcriptional potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MSK1 (Ser376)
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 2F3
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rps6ka5
Gene Accession No. O75582
Gene Alias 3110005L17Rik; 6330404E13Rik; 90 kDa ribosomal protein S6 kinase 5; AI854034; kinase MSK1; KS6A5; mitogen and stress-activated protein kinase 1; Msk1; MSPK1; Nuclear mitogen- and stress-activated protein kinase 1; ribosomal protein S6 kinase A5; ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; ribosomal protein S6 kinase, polypeptide 5; RLPK; RLSK; Rps6ka5; RSKL; RSK-like protein kinase; S6K-alpha-5
Gene Symbols Rps6ka5
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-RPS6KA5 (S376).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9252
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.