Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-mTOR (Ser2448) Recombinant Rabbit Monoclonal Antibody (3L18), Proteintech

Rabbit Recombinant Monoclonal Antibody
Supplier: Proteintech Group Inc 805961RR100UL
Description
Phospho-mTOR (Ser2448)Recombinant Monoclonal antibody specifically detects Phospho-mTOR (Ser2448) in Human, Rat samples. It is validated for Western BlotSpecifications
| Phospho-mTOR (Ser2448) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FRAP; FRAP1; FRAP2; MTOR; RAFT1; RAPT1; SKS | |
| Protein A | |
| RUO | |
| 2475 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 3L18 | |
| Unconjugated | |
| P42345 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction