Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-mTOR (Ser2448) Recombinant Rabbit Monoclonal Antibody (mTORS2448-E11), FITC, Invitrogen™

Catalog No. PIMA537392
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537392 10 Tests
PIMA537146 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537392 Supplier Invitrogen™ Supplier No. MA537392
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-mTOR (Ser2448)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone mTORS2448-E11
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MTOR
Gene Accession No. P42345, Q9JLN9
Gene Alias 2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
Gene Symbols MTOR
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser2448 of human phospho mTOR.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2475, 56717
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.