Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MUNC18 (Ser313) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538296
Description
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Specifications
Phospho-MUNC18 (Ser313) | |
Polyclonal | |
Unconjugated | |
STXBP1 | |
STXBP1, MUNC18-1, UNC18, hUNC18, syntaxin binding protein 1, Hunc18, rbSec1, syntaxin-binding protein 1, RP11-56D16.3, EIEE4, FLJ37475, RBSEC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20910, 25558, 6812 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61764, O08599, P61765 | |
STXBP1 | |
A synthetic phosphopeptide derived from human MUNC-18a around the phosphorylation site of Ser313 (S-S-SP-K-R) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction