Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MYL2 (Tyr118) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567499 Supplier Invitrogen™ Supplier No. PA567499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-MYL2 (Tyr118) Polyclonal Antibody for Western Blot, ICC/IF

Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MYL2 (Tyr118)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYL2
Gene Accession No. P10916
Gene Alias cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mlc2b; MLC-2s/v; MLC2V; MLC-2v; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848
Gene Symbols MYL2
Host Species Rabbit
Immunogen Synthesized peptide sequence derived from Human MYL2 around phosphorylation site of tyrosine 118 (A-D-Y(p)-V-R).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4633
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.