Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MYL6 (Tyr29, Ser30) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501986596
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6596 Supplier Bioss Supplier No. BS5478R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain that is expressed in smooth muscle and non-muscle tissues. Genomic sequences representing several pseudogenes have been described and two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq, Jul 2008].

Specifications

Antigen Phospho-MYL6 (Tyr29, Ser30)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYL6
Gene Accession No. P60660, Q60605, Q64119
Gene Alias 17 kDa myosin light chain; ESMLC; LC17; LC17A; LC17B; LC17-GI; LC17-NM; MLC1SM; MLC-3; MLC3NM; MLC3SM; Myl6; Myln; Myosin light chain 3; myosin light chain 6; Myosin light chain A3; Myosin light chain alkali 3; myosin light chain, alkali, nonmuscle; Myosin light polypeptide 6; myosin, light chain 6, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6, alkali, smooth muscle and non-muscle; Smooth muscle and nonmuscle myosin light chain alkali 6
Gene Symbols MYL6
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MYL6 around the phosphorylation site of Tyr29+Ser30.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17904, 4637, 685867
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.