Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MYL9 (Ser20) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7052R

Description
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.Specifications
Phospho-MYL9 (Ser20) | |
Polyclonal | |
Unconjugated | |
MYL9 | |
20 kDa myosin light chain; AI327049; BOS_13338; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; LOC100343384; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; MYL9; myl9.L; myl9b; myl9l; Mylc2c; myosin; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 12A, regulatory, non-sarcomeric-like; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; regulatory; RLC-C; smooth muscle major isoform; XELAEV_18003093mg; zgc:77916 | |
Rabbit | |
Protein A | |
RUO | |
10398 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P24844 | |
MYL9 | |
KLH conjugated synthetic phosphopeptide derived from human Myosin Regulatory Light polypeptide 9 around the phosphorylation site of Ser20. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction