Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MYL9 (Thr19) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5871 Supplier Bioss Supplier No. BS4060R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.

Specifications

Antigen Phospho-MYL9 (Thr19)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYL9
Gene Accession No. P24844
Gene Alias 20 kDa myosin light chain; AI327049; BOS_13338; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; LOC100343384; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; MYL9; myl9.L; myl9b; myl9l; Mylc2c; myosin; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 12A, regulatory, non-sarcomeric-like; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; regulatory; RLC-C; smooth muscle major isoform; XELAEV_18003093mg; zgc:77916
Gene Symbols MYL9
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MYL9 around the phosphorylation site of Thr19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10398
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.