Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MYOD (Thr115) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105354
Description
Antibody detects endogenous levels of MyoD1 only when phosphorylated at Thr115.
The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).
Specifications
| Phospho-MYOD (Thr115) | |
| Polyclonal | |
| Unconjugated | |
| MYOD1 | |
| AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 17927, 337868, 4654 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10085, P15172, Q02346 | |
| MYOD1 | |
| A synthesized peptide derived from human MYOD1(Accession P15172), corresponding to amino acid residues around phosphorylated Thr115. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction