Learn More
Phospho-Myosin (Ser19, Ser20)Rabbit anti-Human, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 100401416

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Myosin pS19/pS20 antibody is directed against the regulatory light chain of smooth and non-muscle myosin. Reactivity with non-phosphorylated myosin light chain is less than 1% by ELISA. Cross reactivity is expected with myosin light chain from human and mouse. Reactivity with the protein from other species has not been determined. However, the sequence of the immunogen is nearly identical in mammalian and avian species. BLAST search analysis was used to determine that the smooth and non-muscle forms of myosin regulatory light chain have identical sequences. Cross reactivity is expected.
Myosin regulatory light polypeptide 9 is a highly conserved, ubiquitously expressed protein that in humans is encoded by the MYL9 gene. Myosin, a structural component of muscle, are hexameric proteins consisting of two heavy chains and four light chains, including a single pair of nonphosphorylatable light chain subunits and another pair of phosphorylatable light chain subunits.The protein encoded by this gene is a myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. Myosin light chain (Myl) 9 and Myl12 are new functional ligands for CD69. Blockade of CD69-Myl9/12 interactio ameliorates allergic airway inflammation in ovalbumin-induced and house dust mite-induced mouse models of asthma.Specifications
Phospho-Myosin (Ser19, Ser20) | |
Polyclonal | |
Unconjugated | |
MYL12B | |
1500001M02Rik; 20 kDa myosin light chain; 2900073G15Rik; AI327049; AI838292; brain specific myosin regulatory light chain; C77744; C80200; Epididymis secretory protein Li 24; HEL-S-24; LC20; MLC2; MLC-2; MLC20; MLC-2A; MLC-2B; MLC-2C; MLCB; MLC-B; MRCL3; MRLC1; MRLC2; MRLC3; Mrlcb; MYL12A; Myl12b; MYL2B; MYL9; Mylc2b; Mylc2c; myosin light chain 12A; myosin light chain 12B; myosin light chain 9; myosin light chain, regulatory B; myosin light chain, regulatory B-like; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 12A; Myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 20 kDa; Myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain 3; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; Myosin regulatory light chain RLC-A; myosin regulatory light chain-B; Myosin regulatory light polypeptide 9; Myosin RLC; Myosin RLC-A; Myosin RLC-B; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light chain 12B, regulatory; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); Myrl2; NMDA receptor-interacting protein; RGD1309537; RLC; Rlc-a; RLC-B; RLC-C; SHUJUN-1 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
70 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
O14950, P19105, P24844 | |
MYL12A, MYL12B, MYL9 | |
Human Myosin Light Chain phospho peptide corresponding to a region near the amino terminus of the human smooth/non-muscle form of myosin regulatory light chain conjugated to Keyhole Limpet Hemocyanin (KLH). | |
RUO | |
103910, 10398, 10627 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.