Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MYPT1 (Ser507) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106148 Supplier Invitrogen™ Supplier No. PA5106148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MYPT1 only when phosphorylated at Ser507.

Myosin phosphatase target subunit 1, which is also called the myosin-binding subunit of myosin phosphatase, is one of the subunits of myosin phosphatase. Myosin phosphatase regulates the interaction of actin and myosin downstream of the guanosine triphosphatase Rho. The small guanosine triphosphatase Rho is implicated in myosin light chain (MLC) phosphorylation, which results in contraction of smooth muscle and interaction of actin and myosin in nonmuscle cells. The guanosine triphosphate (GTP)-bound, active form of RhoA (GTP. RhoA) specifically interacted with the myosin-binding subunit (MBS) of myosin phosphatase, which regulates the extent of phosphorylation of MLC. Rho-associated kinase (Rho-kinase), which is activated by GTP. RhoA, phosphorylated MBS and consequently inactivated myosin phosphatase. Overexpression of RhoA or activated RhoA in NIH 3T3 cells increased phosphorylation of MBS and MLC. Thus, Rho appears to inhibit myosin phosphatase through the action of Rho-kinase. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MYPT1 (Ser507)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R12A
Gene Accession No. O14974, Q10728, Q9DBR7
Gene Alias 1200015F06Rik; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase (M130); 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase; 130 kDa regulatory subunit of myosin phosphatase; 133kDa myosin-binding subunit of smooth muscle myosin phosphatase (M133); 5730577I22Rik; AA792106; AV099298; D10Ertd625e; hypothetical protein; I79_016312; LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 12A; M110; M130; M130 of smooth muscle myosin phosphatase; MBS; MBSP; myosin binding subunit; myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; myosin phosphatase-targeting subunit 1; myosin-binding subunit of myosin phosphatase; Mypt1; PP-1M; PP1M M110 subunit; PP1M subunit M110; Ppp1r12a; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1M 110 kda regulatory subunit; protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; si:dkey-28j4.1; smooth muscle myosin phosphatase myosin-binding subunit; unm p82emcf; unm_p82emcf; zgc:110448; zgc:110448 protein
Gene Symbols PPP1R12A
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R12A(Accession O14974), corresponding to amino acid residues around phosphorylated Ser507.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116670, 17931, 4659
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.