Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MYPT1 (Thr853) Polyclonal Antibody

Supplier: Thermo Scientific PA5117228
Description
Specifications
| Phospho-MYPT1 (Thr853) | |
| Polyclonal | |
| Unconjugated | |
| PPP1R12A | |
| 1200015F06Rik; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase (M130); 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase; 130 kDa regulatory subunit of myosin phosphatase; 133kDa myosin-binding subunit of smooth muscle myosin phosphatase (M133); 5730577I22Rik; AA792106; AV099298; D10Ertd625e; hypothetical protein; I79_016312; LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 12A; M110; M130; M130 of smooth muscle myosin phosphatase; MBS; MBSP; myosin binding subunit; myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; myosin phosphatase-targeting subunit 1; myosin-binding subunit of myosin phosphatase; Mypt1; PP-1M; PP1M M110 subunit; PP1M subunit M110; Ppp1r12a; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1M 110 kda regulatory subunit; protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; si:dkey-28j4.1; smooth muscle myosin phosphatase myosin-binding subunit; unm p82emcf; unm_p82emcf; zgc:110448; zgc:110448 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116670, 17931, 4659 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O14974, Q10728, Q9DBR7 | |
| PPP1R12A | |
| A synthetic phosphorylated peptide around T853 of human PPP1R12A (NP_0024711). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction