Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PKMYT1 (Ser83) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537656 Supplier Invitrogen™ Supplier No. PA537656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for IHC is human breast carcinoma.

PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKMYT1 (Ser83)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pkmyt1
Gene Accession No. Q99640
Gene Alias 6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg
Gene Symbols Pkmyt1
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 83(R-V-S(p)-F-R) derived from Human MYT1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9088
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.