Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Myt1 (Ser83) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985285
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5285 Supplier Bioss Supplier No. BS3296R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.

Specifications

Antigen Phospho-Myt1 (Ser83)
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYT1
Gene Accession No. Q01538
Gene Alias BOS_13543; C20orf36; KIAA0835; KIAA1050; MTF1; Myelin transcription factor 1; Myelin transcription factor I; MYT1; MYTI; Neural zinc finger factor 2; neural zinc finger transcription factor 2; Nzf2; NZF-2; Nzf2a; Nzf2b; Nztf2; PLPB1; proteolipid protein binding protein; Proteolipid protein-binding protein; ZC2H2C1; ZC2HC4A
Gene Symbols MYT1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Myt1 around the phosphorylation site of Ser83.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4661
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.