Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NBS1 (Ser343) Recombinant Rabbit Monoclonal Antibody (1B4)
GREENER_CHOICE

Catalog No. PIMA533184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533184 Supplier Invitrogen™ Supplier No. MA533184
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-NBS1 (Ser343) Recombinant Monoclonal Antibody for Western Blot, ELISA

The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NBS1 (Ser343)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1B4
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NBN
Gene Accession No. O60934
Gene Alias AT V1; AT V2; ATV; AT-V1; AT-V2; Cell cycle regulatory protein p95; FLJ10155; I79_012607; MGC87362; Nbn; NBS; NBS1; nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; OTTHUMP00000204088; P95; p95 protein of the MRE11/RAD50 complex; p95-NBS1
Gene Symbols NBN
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-NBN (S343).
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4683
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.