Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-NCF1/p47 PHOX (S345) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABF1809100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF1809100 100 μg
ABF180925UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABF1809100 Supplier MilliporeSigma Supplier No. ABF1809100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho-NCF1/p47 PHOX (S345) Polyclonal antibody specifically detects phospho-NCF1/p47 PHOX (S345) in Human samples. It is validated for Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-NCF1/p47 PHOX (S345)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 10 mM Tris-HCl pH7.5 with 0.01% sodium azide with 10% glycerol.
Host Species Rabbit
Immunogen Ovalbumin-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human Neutrophil cytosol factor 1 (NCF1) phosphorylated on serine 345.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.