Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NDRG1 (Thr346) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597401 Supplier Invitrogen™ Supplier No. PA597401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHTSE; Positive Samples: Jurkat; Cellular Location: Cell membrane, Cytoplasm, Nucleus, centrosome, cytoskeleton, cytosol, microtubule organizing center.

NDRG1 is a 394 amino acid cytoplasmic protein belonging to the NDRG family. It interacts with APO A-I and A-II and plays a role in HDL-mediated cholesterol transport, stress responses, hormone responses, cell growth and differentiation. Its defect causes Charcot-Marie-Tooth disease type 4D (CMT4D) that is characterized by reduced nerve conduction velocities, segmental demyelination and remyelination with onion bulb formations on nerve biopsy, slowly progressive distal muscle atrophy and weakness, absent deep tendon reflexes and hollow feet. NDRG1 plays a growth inhibitory role and serves as a marker of differentiation. It is expressed in placental membranes, prostate, kidney, small intestine, ovary tissues, colon and the cells that border the lumen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NDRG1 (Thr346)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NDRG1
Gene Accession No. Q62433, Q6JE36, Q92597
Gene Alias CAP43; CMT4D; differentiation-related gene 1 protein; DRG1; DRG-1; GC4; HMSNL; Ndr1; Ndrg1; Ndrl; nickel-specific induction protein Cap43; NMSL; N-myc downstream regulated 1; N-myc downstream regulated gene 1; N-myc downstream-regulated gene 1 protein; Protein Ndr1; protein NDRG1; protein regulated by oxygen-1; PROXY1; Reducing agents and tunicamycin-responsive protein; Rit42; RTP; TARG1; TDD5
Gene Symbols NDRG1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T346 of human NDRG1 (NP_0011287141).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10397, 17988, 299923
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.