Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NF-kB p100/p52 (Ser865) Polyclonal Antibody, Abbomax

Catalog No. 502203577
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3577 Supplier Abbomax Inc Supplier No. 50011874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: all human tissue Cellular location: Nucleus, Cytoplasm.

Phospho-NF-kB p100/p52 (Ser865) Polyclonal antibody specifically detects Phospho-NF-kB p100/p52 (Ser865) in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen Phospho-NF-kB p100/p52 (Ser865)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene Nfkb2
Gene Accession No. Q00653, Q9WTK5
Gene Alias CVID10; DNA-binding factor KBF2; H2TF1; Lymphocyte translocation chromosome 10 protein; lyt; LYT10; LYT-10; NF kappaB2; NF KB; NF-kappaB2; nfkb p100; NFKB, p52/p100 subunit; NFKB2; NF-kB2; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit 2; nuclear factor Kappa-B, subunit 2; nuclear factor NF-kappa-B p100 subunit; Nuclear factor NF-kappa-B p52 subunit; nuclear factor of Kappa light chain gene enhancer in B cells 2; nuclear factor of kappa light polypeptide gene enhancer in B cells 2, p49/p100; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100); nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, p49/p100; oncogene Lyt-10; p100; p49; p49/p100; p50B; p52; RGD1307189; transcription factor NFKB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the epitope containing a phosphorylation site Ser 865 of human NFkB/p100/p52 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18034, 4791
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.