Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NF2 (Ser10) Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA540266

Catalog No. PA540266


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-NF2 (Ser10)
Polyclonal
Unconjugated
Nf2
ACN; BANF; Merlin; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); neurofibromin 2b (merlin); neurofibromin-2; Nf2; Nf-2; NF2a; nf2b; OTTHUMP00000199607; SCH; Schwannomerlin; schwannomin
Rabbit
Antigen affinity chromatography
RUO
18016, 25744, 4771
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P35240, P46662, Q63648
Nf2
Synthesized peptide derived from human NF2 around the phosphorylation site of S10.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.