Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-NF2 (Ser518) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517072
Description
It is not recommended to aliquot this antibody.
NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.
Specifications
Phospho-NF2 (Ser518) | |
Polyclonal | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
P35240, P46662, Q63648 | |
Nf2 | |
Synthetic peptide corresponding to the sequence of human merlin. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Nf2 | |
ACN; BANF; Merlin; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); neurofibromin 2b (merlin); neurofibromin-2; Nf2; Nf-2; NF2a; nf2b; OTTHUMP00000199607; SCH; Schwannomerlin; schwannomin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18016, 25744, 4771 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction