Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NFAT5 (Ser1197) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA538884

Catalog No. PIPA538884


Description

Description

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-NFAT5 (Ser1197)
Polyclonal
Unconjugated
NFAT5
Nuclear Factor Activated T-Cells, NFATL1, Nuclear factor of activated T cells 5, T cell transcription factor NFAT5, NF-AT5, Tonicity-responsive enhanc
Rabbit
Antigen affinity chromatography
RUO
10725, 54446
-20°C
Liquid
Peptide ELISA, Immunofluorescence
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
O94916, Q9WV30
NFAT5
A synthetic phosphopeptide derived from human NFAT5 around the phosphorylation site of Ser1197 (M-L-SP-Q-E)
100 μg
Primary
Human, Mouse, Non-human Primate
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.