Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NFAT5 (Ser145) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105436 Supplier Invitrogen™ Supplier No. PA5105436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of NFAT5 only when phosphorylated at Ser145.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NFAT5 (Ser145)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NFAT5
Gene Accession No. O94916, Q9WV30
Gene Alias AI225870; B130038B15Rik; CAG-8; CAG80; glutamine rich protein H65; KIAA0827; mKIAA0827; Nfat; Nfat5; NF-AT5; NFATL1; NFAT-like protein 1; nfatz; nuclear factor of activated T cells 5; nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; OREBP; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; TONEBP; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein
Gene Symbols NFAT5
Host Species Rabbit
Immunogen A synthesized peptide derived from human NFAT5(Accession O94916), corresponding to amino acid residues around phosphorylated Ser145.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10725, 54446
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.