Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NFATC1 (Ser294) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538301
Description
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
Specifications
Phospho-NFATC1 (Ser294) | |
Polyclonal | |
Unconjugated | |
NFATC1 | |
NF-ATC, NFAT2, NFATc, NF-ATc1, NFAT transcription complex cytosolic component, nuclear factor of activated T-cells, cytoplasmic 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18018, 4772 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95644, O88942 | |
NFATC1 | |
A synthetic phosphopeptide derived from human NFAT2 around the phosphorylation site of Ser294 (R-V-SP-V-T) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction