Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NFATC2 (Ser326) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105754 Supplier Invitrogen™ Supplier No. PA5105754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of NFAT1 only when phosphorylated at Ser326.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NFATC2 (Ser326)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NFATC2
Gene Accession No. Q13469, Q60591
Gene Alias AI607462; IB-IXL; KIAA0611; NFAT pre-existing s; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; Nfat1; NFAT1-D; Nfatc2; NF-ATc2; Nfatp; NF-ATp; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2; nuclear factor of activated T-cells 2; nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; OTTHUMP00000031291; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Symbols NFATC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NFATC2(Accession Q13469), corresponding to amino acid residues around phosphorylated Ser326.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18019, 311658, 4773
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.