Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NFATC3 (Ser165) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538303
Description
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
Specifications
Phospho-NFATC3 (Ser165) | |
Polyclonal | |
Unconjugated | |
NFATC3 | |
nuclear factor of activated T-cells cytoplasmic calcineurin-dependent 3, NFATX, T cell transcription factor NFAT4, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells cytoplasmic 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18021, 4775 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q12968, P97305 | |
NFATC3 | |
A synthetic phosphopeptide derived from human NFAT4 around the phosphorylation site of Ser165 (S-P-SP-P-A) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction