Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-NFATC4 (Ser289) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105650
Description
Antibody detects endogenous levels of NFAT3 only when phosphorylated at Ser289.
This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.
Specifications
| Phospho-NFATC4 (Ser289) | |
| Polyclonal | |
| Unconjugated | |
| Nfatc4 | |
| 3110041H08Rik; AW107667; AW546455; Nfat3; NF-AT3; Nfatc4; NF-ATc4; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4; nuclear factor of activated T-cells 4; nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 305897, 4776, 73181 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14934, Q8K120 | |
| Nfatc4 | |
| A synthesized peptide derived from human NFATC4(Accession Q14934), corresponding to amino acid residues around phosphorylated Ser289. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction