Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NFkB-p100 (Ser866) Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278049
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8049 Supplier Leinco Technologies Inc Supplier No. 43015100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-NFkB-p100 (Ser866) Polyclonal antibody specifically detects Phospho-NFkB-p100 (Ser866) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen Phospho-NFkB-p100 (Ser866)
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco's PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nfkb2
Gene Accession No. Q00653, Q9WTK5
Gene Alias CVID10; DNA-binding factor KBF2; H2TF1; Lymphocyte translocation chromosome 10 protein; lyt; Lyt10; LYT-10; NF kappaB2; NF-kappaB2; NFKB, p52/p100 subunit; NFKB2; NF-kB2; nuclear factor kappa B subunit 2; nuclear factor Kappa-B, subunit 2; nuclear factor NF-kappa-B p100 subunit; Nuclear factor NF-kappa-B p52 subunit; nuclear factor of Kappa light chain gene enhancer in B cells 2; nuclear factor of kappa light polypeptide gene enhancer in B cells 2, p49/p100; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100); nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, p49/p100; oncogene Lyt-10; p100; p49; p49/p100; p50B; p52; RGD1307189; transcription factor NFKB2
Host Species Rabbit
Immunogen Peptide sequence that includes phosphorylation site of Serine 866 (E-D-S(p)-A-Y) derived from human NF kappa B-p100 and conjugated to KLH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18034, 309452, 4791
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.