Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NFkB-p65 (Ser276) Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
Supplier: Leinco Technologies Inc 43011100UL
This item is not returnable.
View return policy
Description
Phospho-NFkB-p65 (Ser276) Polyclonal antibody specifically detects Phospho-NFkB-p65 (Ser276) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western BlotSpecifications
| Phospho-NFkB-p65 (Ser276) | |
| Polyclonal | |
| Unconjugated | |
| RELA | |
| avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; Xrel1; xrela | |
| Peptide sequence that includes phosphorylation site of Serine 276 (R-P-S(p)-D-R) derived from human NF kappa B-p65 and conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Dulbecco's PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q04206, Q04207 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19697, 309165, 5970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction