Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NFkB p65 (Ser529) Recombinant Rabbit Monoclonal Antibody (NFkBp65S529-H3), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$408.50 - $774.00
Specifications
| Antigen | Phospho-NFkB p65 (Ser529) |
|---|---|
| Clone | NFkBp65S529-H3 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA537411
|
Invitrogen™
MA537411 |
20 μL |
Each for $408.50
|
|
|||||
PIMA537164
![]() |
Invitrogen™
MA537164 |
200 μL |
Each for $774.00
|
|
|||||
Description
Nuclear factor kB p65 (NF-kB p65) is encoded by the RELA gene and is present on chromosome 11 in humans. NF-kB P65 is also known as RelA (v-rel avian reticuloendotheliosis viral oncogene homolog A) and belongs to the Rel family of proteins. It is one of the two subunits of NF-kB (Nuclear factor kappa-light-chain-enhancer of activated B cells) that heterodimerizes with the other subunits p50 or p52. However, binding of TNF alpha to its cognate receptor phosphorylates IKK which in turn phosphorylates I kB allowing proteasomal degradation of I kB. It specifically plays a key role in transcription of immunoglobulin k (kappa) gene in mature B-lymphoid cells.Specifications
| Phospho-NFkB p65 (Ser529) | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
| avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; Xrel1; xrela | |
| RELA | |
| IgG κ | |
| Protein A/G | |
| Antibody |
| NFkBp65S529-H3 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q04206, Q04207 | |
| 19697, 5970 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser529 of human phospho-NFKB p65. | |
| Primary | |
| -20°C | |
| RELA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title