Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active Motif Phospho-NFkB p65 (Ser536) Polyclonal Antibody
Rabbit Polyclonal Antibody
Specifications
Antigen | Phospho-NFkB p65 (Ser536) |
---|---|
Concentration | Conc. Not Determined |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
SPN-39675
|
100 μL | N/A | N/A | N/A | |||||
Description
Phospho-NFkB p65 (Ser536) Polyclonal Antibody for Western Blot, DB
Nuclear factor kB p65 (NF-kB p65) is encoded by the RELA gene and is present on chromosome 11 in humans. NF-kB P65 is also known as RelA (v-rel avian reticuloendotheliosis viral oncogene homolog A) and belongs to the Rel family of proteins. It is one of the two subunits of NF-kB (Nuclear factor kappa-light-chain-enhancer of activated B cells) that heterodimerizes with the other subunits p50 or p52. However, binding of TNF alpha to its cognate receptor phosphorylates IKK which in turn phosphorylates I kB allowing proteasomal degradation of I kB. It specifically plays a key role in transcription of immunoglobulin k (kappa) gene in mature B-lymphoid cells.Specifications
Phospho-NFkB p65 (Ser536) | |
-20°C, Avoid Freeze/Thaw Cycles | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
RELA | |
avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; Xrel1; xrela | |
RELA | |
Primary |
Conc. Not Determined | |
Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
Whole serum with 30% glycerol and 0.035% sodium azide | |
Q04206 | |
5970 | |
This NFkB p65 phospho Ser536 antibody was raised against a peptide containing phospho-serine 536 of human NFkB p65. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title