Learn More
Phospho-NFkB (RelA) (Ser276)Rabbit anti-Human, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 100401264
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This phospho specific polyclonal antibody is specific for phosphorylated pS276 human p65. Reactivity with non-phosphorylated p65 is minimal. Cross reactivity with pS276 phosphorylated p65 from mouse, rat or other species has not been determined.
Nuclear factor kB p65 (NF-kB p65) is encoded by the RELA gene and is present on chromosome 11 in humans. NF-kB P65 is also known as RelA (v-rel avian reticuloendotheliosis viral oncogene homolog A) and belongs to the Rel family of proteins. It is one of the two subunits of NF-kB (Nuclear factor kappa-light-chain-enhancer of activated B cells) that heterodimerizes with the other subunits p50 or p52. However, binding of TNF alpha to its cognate receptor phosphorylates IKK which in turn phosphorylates I kB allowing proteasomal degradation of I kB. Phosphorylation of p65 at Ser 536 located in the carboxy-terminal transactivation domains (TAD) leads to nuclear localization of the transcriptionally active complex and NF-kB mediated transactivation of several downstream genes. It specifically plays a key role in transcription of immunoglobulin k (kappa) gene in mature B-lymphoid cells.Specifications
| Phospho-NFkB (RelA) (Ser276) | |
| Polyclonal | |
| Unconjugated | |
| RELA | |
| avian reticuloendotheliosis viral (v-rel) oncogene homolog A; BOS_24973; EBP-1; I79_021812; MGC131774; NFkappaB p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NFKB1; Nfkb3; NFkB-p50; nuclear factor kappa B subunit p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; Putative transcription factor p65 homolog; rel1; Rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; XRel1; xrela | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 80 mg/mL | |
| Antibody with 0.01% sodium azide; pH | |
| Q04206 | |
| RELA | |
| NFkB p65 (Rel A) peptide corresponding to an internal region near phospho Serine 276 of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| RUO | |
| 5970 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.