Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-NHE3 (Ser605) Monoclonal Antibody (10A8), DyLight™ 488
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516089
Description
Suggested positive control: PKA treated rat kidney membrane.
NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
Specifications
Phospho-NHE3 (Ser605) | |
Monoclonal | |
2.1 mg/mL | |
50mM sodium borate with no preservative | |
G3X939, P26432, P26433 | |
SLC9A3 | |
Synthetic peptide made to a region surrounding the Serine 605 portion of the rat protein. | |
100 μL | |
Primary | |
Mouse, Rat, Rabbit, Monkey, Pig, Marsupial | |
Antibody | |
IgG2b |
Western Blot, Immunocytochemistry | |
10A8 | |
DyLight 488 | |
SLC9A3 | |
9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 | |
Mouse | |
Protein G | |
RUO | |
100009430, 105243, 24784, 397358 | |
4°C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction