Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NHE3 (Ser605) Monoclonal Antibody (10A8), DyLight™ 650
GREENER_CHOICE

Catalog No. PIMA516088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516088 Supplier Invitrogen™ Supplier No. MA516088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: PKA treated rat kidney membrane.

NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NHE3 (Ser605)
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10A8
Concentration 2.1 mg/mL
Conjugate DyLight 650
Formulation 50mM sodium borate with 0.05% sodium azide
Gene SLC9A3
Gene Accession No. G3X939, P26432, P26433
Gene Alias 9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Gene Symbols SLC9A3
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009430, 105243, 24784, 397358
Target Species Mouse, Rat, Rabbit, Monkey, Pig, Marsupial
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.