Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Niban-like protein (Ser679, Ser683) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595683 Supplier Invitrogen™ Supplier No. PA595683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antigen Affinity Purified from Pooled Serum.

FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Niban-like protein (Ser679, Ser683)
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene Fam129b
Gene Accession No. Q8R1F1, Q96TA1
Gene Alias 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018
Gene Symbols Fam129b
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser679/683 conjugated to KLH.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227737, 64855
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.