Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NLRC4 (Ser533) Monoclonal Antibody (M549)
GREENER_CHOICE

Catalog No. PIMA547631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547631 Supplier Invitrogen™ Supplier No. MA547631
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This antibody antibody detects a 110 kDa* protein on SDS-PAGE immunoblots of PMA-differentiated THP1 human macrophages. In addition, the antibody detects a mouse recombinant phospho-NLRC4 (Ser-533) peptide, but does not detect this peptide after lambda phosphatase treatment.

NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NLRC4 (Ser533)
Applications Western Blot
Classification Monoclonal
Clone M549
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Nlrc4
Gene Accession No. F1M649, Q3UP24, Q9NPP4
Gene Alias 9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215
Gene Symbols Nlrc4
Host Species Rat
Immunogen Clone M549 was generated from a phospho-NLRC4 (Ser-533) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding serine 533 in mouse NLRC4. This sequence has high homology to human and rat NLRC4, and is not homologous to sequences in other NLR family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268973, 298784, 58484
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.