Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NMDAR2A (Tyr943) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105627 Supplier Invitrogen™ Supplier No. PA5105627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of NMDAR2A only when phosphorylated at Tyr943.

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NMDAR2A (Tyr943)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Grin2a
Gene Accession No. P35436, Q00959, Q12879
Gene Alias EPND; FESD; GluN2A; GluRepsilon1; glutamate [NMDA] receptor subunit epsilon-1; glutamate ionotropic receptor NMDA type subunit 2A; glutamate receptor; glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, NMDA2A (epsilon 1); glutamate receptor, ionotropic, N-methyl D-aspartate 2A; Grin2a; hNR2A; LKS; NMDA Receptor 2A; NMDAR 2A; NMDAR2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NR2A; OTTHUMP00000160135; OTTHUMP00000174531
Gene Symbols Grin2a
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRIN2A(Accession Q12879), corresponding to amino acid residues around phosphorylated Tyr943.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14811, 24409, 2903
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.