Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NMDAR2B (Tyr1474) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537591 Supplier Invitrogen™ Supplier No. PA537591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is A431 cells; suggested positive control for ICC/IF is Hela cells.

NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NMDAR2B (Tyr1474)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRIN2B
Gene Accession No. Q00960, Q01097, Q13224
Gene Alias AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3
Gene Symbols GRIN2B
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Tyr1474 (H-V-Y(p)-E-K) derived from Human NMDAR2B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14812, 24410, 2904
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.