Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-NR5A1 (Ser203) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106044 Supplier Invitrogen™ Supplier No. PA5106044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Steroidogenic Factor 1 only when phosphorylated at Ser203.

Steroidogenic factor-1 (SF-1) is an orphan nuclear receptor involved in organ development and differentiation of the endocrine system. SF-1 has been shown to be essential for fetal development of the hypothalamus, pituitary gland, gonads, and the adrenal gland. SF-1 has been shown to promote transcriptional activity of many steroidogenic enzymes from the cytochrome P450 family. SF-1 contains two activation function domains, designated AF1 and AF2, common to many nuclear steroid receptors which have been shown to be necessary for interaction with other nuclear receptors as well as coactivators and corepressors including steroid receptor coactivator-1 (SRC-1), glucocorticoid receptor interacting protein 1 (GRIP1), cAMP response element binding protein (CREB) binding protein (CBP) and silencing mediator of retinoid acid and thyroid hormone receptor (SMRT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NR5A1 (Ser203)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nr5a1
Gene Accession No. P33242, P50569, Q13285
Gene Alias Ad4BP; Ad4BP/SF-1; adrenal 4 binding protein; Adrenal 4 binding protein (AD4BP); adrenal 4-binding protein; ELP; ELP-3; Embryonal long terminal repeat-binding protein; embryonal LTR-binding protein; FTZ1; FTZF1; Ftz-F1; fushi tarazu 1 factor homolog; fushi tarazu factor homolog 1; Fushi tarazu factor homolog 1 (FTZ1); hSF-1; Nr5a1; nuclear receptor AdBP4; nuclear receptor subfamily 5 group A member 1; Nuclear receptor subfamily 5 group A member 1 (NR5A1); nuclear receptor subfamily 5, group A, member 1; POF7; SF1; SF-1; SPGF8; SRXY3; Steroid hormone receptor Ad4BP; steroid hydroxylase positive regulator; Steroidogenic factor 1; Steroidogenic factor 1 (SF1); steroidogenic factor 1 nuclear receptor; steroidogenic factor-1; STF1; STF-1
Gene Symbols Nr5a1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NR5A1(Accession Q13285), corresponding to amino acid residues around phosphorylated Ser203.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2516, 26423, 83826
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.