Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NT5C (Ser184) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105680 Supplier Thermo Scientific Supplier No. PA5105680
Add to Cart
Add to Cart

Phospho-NT5C (Ser184) Polyclonal Antibody

Phospho-NT5C (Ser184) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-NT5C (Ser184)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NT5C
Gene Accession No. Q8TCD5, Q9JM14
Gene Alias 5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; 5', 3'-nucleotidase, cytosolic; 5',3'-nucleotidase, cytosolic; 5,3-nucleotidase, cytosolic; cdN; Cytosolic 5',3'-pyrimidine nucleotidase; deoxy-5'-nucleotidase 1; Dnt; DNT1; dNT-1; epididymis luminal protein 74; HEL74; Nt5c; P5N2; PN-I; PN-II; Umph2; Umph-2; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2; uridine monophosphatase 2
Gene Symbols NT5C
Host Species Rabbit
Immunogen A synthesized peptide derived from human NT5C(Accession Q8TCD5), corresponding to amino acid residues around phosphorylated Ser184.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360582, 30833, 50773
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.