Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-OCT1 (POU2F1) (Ser385) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5105826
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105826 Supplier Thermo Scientific Supplier No. PA5105826

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of POU2F1/OCT1 only when phosphorylated at Ser385.

OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-OCT1 (POU2F1) (Ser385)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POU2F1
Gene Accession No. P14859, P25425, P31503
Gene Alias 2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor
Gene Symbols POU2F1
Host Species Rabbit
Immunogen A synthesized peptide derived from human POU2F1(Accession P14859), corresponding to amino acid residues around phosphorylated Ser385.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171068, 18986, 5451
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.