Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-OPRD1 (Ser363) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4432 Supplier Bioss Supplier No. BS20197R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer′s patients and in the putamen in Parkinson′s disease.

Specifications

Antigen Phospho-OPRD1 (Ser363)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OPRD1
Gene Accession No. P41143
Gene Alias delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; Opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; OPRD1; Ror-a
Gene Symbols OPRD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OPRD1 around the phosphorylation site of TP (p-S)DG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4985
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.