Learn More
Invitrogen™ Phospho-OPRK1 (Ser369) Polyclonal Antibody

Description
Phospho-OPRK1 (Ser369) Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
Specifications
Specifications
| Antigen | Phospho-OPRK1 (Ser369) |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | OPRK1 |
| Gene Accession No. | P33534, P34975 |
| Gene Alias | kappa opioid receptor; kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; OPRK1; Oprk2; Ork1; R21; Ror-d |
| Gene Symbols | OPRK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.