Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-OSR1 (Thr185) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA537804
Description
A suggested positive control for Western blot is HepG2 cells; suggested positive control for IHC is human breast carcinoma.
Phospho-OSR1 (Thr185) Polyclonal antibody specifically detects Phospho-OSR1 (Thr185) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
Phospho-OSR1 (Thr185) | |
Polyclonal | |
Unconjugated | |
Osr1 | |
ODD; Odd1; odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila); odd-skipped related transciption factor 1; odd-skipped related transcription factor 1; Osr; OSR1; Protein odd-skipped-related 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
130497 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8TAX0 | |
Osr1 | |
Peptide sequence around phosphorylation site of threonine185(R-K-T(p)-F-V) derived from Human OSR1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction