Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103022 Supplier Invitrogen™ Supplier No. PA5103022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OSR1.

OSR1 is a transcription factor that plays a role in the regulation of embryonic heart and urogenital development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Osr1
Gene Accession No. Q8TAX0
Gene Alias ODD; Odd1; odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila); odd-skipped related transciption factor 1; odd-skipped related transcription factor 1; Osr; OSR1; Protein odd-skipped-related 1
Gene Symbols Osr1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OSR1, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130497
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.