Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-OXSR1 (Thr185) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103272 Supplier Invitrogen™ Supplier No. PA5103272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of OSR1 only when phosphorylated at Thr185.

OXSR1 serine/threonine kinase or (OSR1) is activated by osmotic stresses and regulates downstream protein kinases such as PAK1 in order to establish the sensor/signaling modules that initiate the cellular response to environmental stress. OXSR1 can be phosphorylated by ATM or ATR during the DNA damage process. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-OXSR1 (Thr185)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Oxsr1
Gene Accession No. O95747, Q6P9R2
Gene Alias 2210022N24Rik; 2810422B09Rik; AI462649; AW209236; C6orf182; CEP57R; KIAA1101; mKIAA1101; Osr1; OTTHUMP00000209062; oxidative stress responsive 1; oxidative stress responsive kinase 1; oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1
Gene Symbols Oxsr1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OXSR1(Accession O95747), corresponding to amino acid residues around phosphorylated Thr185.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108737, 316064, 9943
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.