Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-p130 (Ser952) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105778
Description
Antibody detects endogenous levels of RBL2 only when phosphorylated at Ser952.
RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
Specifications
Phospho-p130 (Ser952) | |
Polyclonal | |
Unconjugated | |
RBL2 | |
130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
19651, 5934, 81758 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O55081, Q08999, Q64700 | |
RBL2 | |
A synthesized peptide derived from human RBL2(Accession Q08999), corresponding to amino acid residues around phosphorylated Ser952. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction