Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GRF-1 (Tyr1105) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105034 Supplier Invitrogen™ Supplier No. PA5105034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GRF-1 only when phosphorylated at Tyrosine 1105.

RNA polymerase (pol) III synthesizes tRNA, 5s rRNA, 7SL RNA and U6 snRNA and is overexpressed in many transformed cell lines and tumors in vivo, since cells must duplicate its protein components before division. Therefore, in order to maintain rapid growth, cells must produce a high level of Pol III transcribed RNA, which requires the presence of the TFIIIB and TFIIIC2 transcription factor complexes. The TFIIIC2 complex is composed of five subunits, TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, that are overexpressed in adenovirus transformed cells as well as in malignant cells in vivo, such as ovarian carcinomas. TFIIIC2 recruits RNA pol III and TFIIIB to promoter elements and may be a key component in the deregulation of malignant cells. The TFIIIB complex includes the TATA-binding protein (TBP), TFIIB-related factor 1 (TFIIIB90, BRF1) and TFIIIB, the expression of which are also upregulated in transformed cells. In many carcinomas, the tumor suppressors retinoblastoma (RB) and p53 are inactivated, which affects their ability to bind and inactivate the function of TFIIIB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GRF-1 (Tyr1105)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP35
Gene Accession No. P81128, Q91YM2, Q9NRY4
Gene Alias 6430596G11Rik; AI841135; ARHGAP35; GAP-associated protein p190; glucocorticoid receptor DNA binding factor 1; Glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF1; GRF-1; GRLF1; KIAA1722; LOW QUALITY PROTEIN: rho GTPase-activating protein 35; MGC10745; mKIAA1722; P190 RhoGAP; P190A; p190-A; p190ARHOGAP; p190RhoGAP; rho GAP p190A; Rho GTPase activating protein 35; rho GTPase-activating protein 35; Rho-GAP protein; p190-A; GRLF1
Gene Symbols ARHGAP35
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP35(Accession Q9NRY4), corresponding to amino acid residues around phosphorylated Tyr1105.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232906, 2909, 306400
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.