Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-p21 (Ser130) Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA512644

Catalog No. PIPA512644


Only null left
Add to Cart

Description

Description

Phospho-p21 (Ser130) Polyclonal Antibody for Western Blot, DB

Protein p21 is a tumor suppressor protein which is an inhibitor of Cyclin-dependent kinases (CDK) and is transcriptionally activated by p53. P21 is important in the response of cells to genotoxic stress and a major transcriptional target of p53 protein. The occurrence of p21 in the nucleus executes binding and inhibition activity of cyclin dependent kinases Cdk1 and Cdk2, and blocks the transition from G1 phase into S phase or from G2 phase into mitosis after DNA damage, which enables the repair of damaged DNA. In the cytoplasm, p21 protein has an anti-apoptotic effect. P21 is able to bind to and inhibit caspase 3, as well as the apoptotic kinases ASK1 and JNK. P21 exhibits a dual function in carcinogenesis, and acts as a tumor suppressor, prevents apoptosis, and acts as an oncogene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-p21 (Ser130)
Polyclonal
Unconjugated
CDKN1A
Activating Fragment 1; CAP20; cdk inhibitor CIP1; cdk inhibitor CIP1 (p21); CDKI; CDK-interacting protein 1; CDK-interaction protein 1; Cdkn1; CDKN1A; CDNK1A; CIP1; Cip1); cyclin dependent kinase inhibitor 1A; cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (P21); cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA6; MDA-6; melanoma differentiation associated protein 6; melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21>WAF1/CIP1>; p21Cip1; p21Cip1/Waf1; p21WAF; PIC1; putative cyclin-dependent kinase inhibitor 1A (p21/WAF1/CIP1); SDI1; SLC12A9; UV96; WAF1; Wild type p53 activated fragment 1 (WAF1); wild-type p53-activated fragment 1
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
1026
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Dot Blot, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P38936
CDKN1A
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S130 of human p21Cip1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.