Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-p23 (Ser113) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538337
Description
Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes.
Specifications
| Phospho-p23 (Ser113) | |
| Polyclonal | |
| Unconjugated | |
| PTGES3 | |
| P23, TEBP, cPGES, Hsp90 co-chaperone, cytosolic prostaglandin E synthase, cytosolic prostaglandin E2 synthase, progesterone receptor complex p23, prostaglandin E synthase 3, telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10728, 362809, 56351 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15185, Q9R0Q7, P83868 | |
| PTGES3 | |
| A synthetic phosphopeptide derived from human TEBP around the phosphorylation site of Ser113 (D-D-SP-D-E) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction