Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-p300 (Ser1834) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564531
Description
Phospho-p300 (Ser1834) Polyclonal Antibody detects endogenous levels of p300 only when phosphorylated at Ser1834.
EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CPB (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CPB can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.
Specifications
Phospho-p300 (Ser1834) | |
Polyclonal | |
Unconjugated | |
EP300 | |
A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170915, 2033, 328572 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
B2RWS6, Q09472 | |
EP300 | |
A synthesized peptide derived from human E1A-associated Protein p300 (Phospho-Ser1834). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction